Learn More
enQuireBio™ Recombinant Rat MMP7 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) MMP7, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6371-ye-500ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_036996.1 | |
MMP7 Protein | |
500 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
25335 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Mmp7 | |
Recombinant Protein | |
Yeast | |
FSLMPNSPKW HSRTVTYRIV SYTTDLPRFL VDQIVKRALR MWSMQIPLNF KRVSWGTADI IIGFARGDHG DNFPFDGPGN TLGHAFAPGP GLGGDAHFDK DEYWTDGEDS GVNFLFVATH ELGHSLGLGH SSVPSSVMYP TYQGDHSEDF SLTKDDIAGI QKLYGKRNKL |