Learn More
enQuireBio™ Recombinant Rat Perforin-1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Perforin-1, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP6527-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
Perforin-1 Protein | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
Greater than 84.2% as determined by SDS-PAGE. | |
Research Use Only | |
PRF1 | |
Recombinant Protein | |
E. coli | |
ATRSECKQNH KFVPGVWAAG EGVDVTTLRR SSSFPVNTGK FLRPDRTCTL CKNALMNDGI QRLPVAIAHW RPHGSHCQRN VATTKVSSTE GVAREAAANI NNDWRAGLDV NPKPEANVHV SVAGSHSKIA NFAAEKAHQD QYNFNTDTVE CRMYSFRLAQ KPPLHPDFRK ALKNLPHNFN SSTEHAYRRL ISSYGTHFIT AVDLGGRVSV LTALRTCQLT LDGLTADEVG DCLSVEAQVS IGAQASVSSE YKACEEKKKQ HKIATSFHQT YRERHVEVLG GPLDSSNDLL FGNQATPEHF STWIASLPTR PDVVDYSLEP LHILLEDSDP KREALRQAIS HYVMSRARWR DCNRPCRAGQ HKSSRDSCQC VCQDSNVTNQ DCCPRQRGLA KLMVRNFQAK GLWGDYITST DAYLKVFFGG QEIRTGVVWN NNHPSWSDKM DFGNVLLSTG GPLRVQVWDA DNGWDDDLLG TCDKSPKSGF HEVNCPLNHG SIKFIYQANC LPDLTGETCL EYAPQGLLGD PRGNRSGAVW |