Learn More
enQuireBio™ Recombinant Rhesus Macaque IL 5 Protein
A cDNA sequence encoding the IL 5 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10717-2ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
710622 | |
Greater than 98.0% as determined by HPLC and SDS-PAGE. | |
Research Use Only | |
IL5 | |
Recombinant Protein | |
E. coli | |
IPTEIPASAL VKETLALLST HRTLLIANET LRIPVPVHKN HQLCTEEIFQ GIGTLESQTV QGGTVERLFK NLSLIKKYIG GQKKKCGEER RRVNQFLDYL QEFLGVMNTE WIIES |
IL 5 Protein | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 5ng/ml, corresponding to a specific activity of > 2.0 M 105 IU/mg. | |
Rhesus Macaque | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in 1xPBS, pH 7.4 and 5% Trehalose. |