missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Zika NS1 Protein

A cDNA sequence encoding the Zika NS1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP14001-HIS-INSECT-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15975379

Quantité Prix
1 171.64€ / 10µg
10 161.27€ / 10µg
Date d’expédition estimée: 25-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

Zika NS1 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Zika
His
Zika NS1 protein solution contains PBS, 25mM arginine and 1M urea.
Zika NS1 protein is >95% pure as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
MKMASNDATP SNDGAAGLVP EINNEAMALD PVAGAAIAAP LTGQQNIIDP WIMNNFVQAP GGEFTVSPRN SPGEVLLNLE LGPEINPYLA HLARMYNGYA GGFEVQVVLA GNAFTAGKII FAAIPPNFPI ENLSAAQITM CPHVIVDVRQ LEPVNLPMPD VRNNFFHYNQ GSDSKLRLVA MLYTPLRANN SGDDVFTVSC RVLTRPSPDF SFNFLVPPTV ESKTKPFTLP ILTISEMSNS RFPVPIDSLH TSPTENIVVQ CQNGRVTLDG ELMGTTQLLP SQICAFRGVL TRSTSRASDQ ADTATPRLFN YYWHIQLDNL NGTPYDPAED IPGPLGTPDF RGKVFGVASQ RNPDSTTRAH EAKVDTTAGR FTPKLGSLEI STESGDFDQN QPTRFTPVGI GVDHEADFQQ WSLPDYSGQF THNMNLAPAV APNFPGEQLL FFRSQLPSSG GRSNGILDCR VPQEWVQHFY QESAPAQTQV ALVRYVNPDT GRVLFEAKLH KLGFMTIAKN GDSPITVPPN GYFRFESWVN PFYTLAPMGT GNGRRRIQ
Vidéos
FDS
Documents
Certificats