missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Congélateurs de laboratoire Revco™ High-Performance

Simplifiez le fonctionnement et améliorez la précision grâce au système à microprocesseur des congélateurs de laboratoire haute performance Thermo Scientific™ Revco™.

Marque:  Thermo Scientific™ ULT430D

Code nomenclature Nacres: NB.84

Informations supplémentaires : Poids : 50.00000kg

 Afficher plus de versions de ce produit

Code produit. 15118135

  • 3932.00€ / 1 pièce


Pour délai, nous consulter 03 88 67 14 14
Ajouter au panier

Description

Description

  • Temperature factory-preset at -20° or -30°C
  • Capacities from 4.9 to 51.1 cu. ft. (139 to 1447L)
  • Icon-based controls and displays
  • Large, easy-to-read digital display shows temperature within 1°C
  • Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature, flashes bulb for undertemperature

Microprocessor Control

  • Audible/visual warnings for overtemperature, undertemperature, or power failure
  • Key-operated OFF/ON/ALARM ON master switch for setpoint security
  • Push-button alarm test
  • Remote alarm contacts
  • Alarm silence, ringback, and auto reset
  • Controller battery backup system

Construction

  • Scratch-resistant cold-rolled steel interior and exterior finishes
  • High-density, 2in. thick (5.1cm) CFC-free urethane foam insulation protects against ambient temperature fluctuations
  • CFC-free, HFC-blend refrigerant
  • Single-door models are hinged on right
  • Full-length door handle and key lock
  • Doors self-close and lock open at 90°
  • Adjustable epoxy-coated wire shelves
  • 1in. (2.5cm) dia. access port with plug on undercounter model

Model Specific Features:

-30°C Freezers

  • Automatic defrost and condensate removal
  • ensor automatically initiates defrost cycle on demand, eliminating need for a defrost timer, minimizing frost buildup, and optimizing coil efficiency
  • Forced-air circulation improves temperature uniformity, provides faster recovery after door is opened

-20°C Freezers

  • Suitable for applications in which slight intermittent coil warming during an automatic defrost cycle is not desired (such as enzyme storage)
  • Gravity convection air system for maximum stability without a blower or fan
  • Manual defrost

Ordering Information:

Alarm systems, temperature chart recorders, and power monitors are available separately.

Note:

Published interior volumes are nominal; usable capacities may be offset by shelves, drawers, blower coils or cabinet protrusions. For specific cabinet dimensions and allowances, or for verification of clearance tolerances, contact your Customer Service Representative.

Spécification

Spécification

UL and CUL listed. CE marked.
28.9 x 24 x 34 in. (73.4 x 61 x 86.4cm)
NEMA 6-15P
High-Performance Freezer
Single, Solid
Warranty varies by country, please contact us for details
15A
86.4cm
60Hz
CFC-free
96kg
50.8cm
Automatic
139L
20.5 x 20 x 20 in. (52.1 x 50.8 x 50.8cm)
−30°C
208 - 230V
208 to 230V 60Hz
6A
50.8cm
52.1cm
73.4cm
3
Microprocessor
Undercounter
61cm
Vidéos