Learn More
ACROBiosystems Rhesus macaque PCSK9 Protein, His Tag
Marque:  ACROBiosystems PC9-C52H2-50UG
Informations supplémentaires : Poids : 0.00001kg
 Afficher plus de versions de ce produit
Code produit. 15867427
  • 262.28€ / 50µg
Date d’expédition estimée: 05-01-2022
Description
Description
Proprotein convertase subtilisin/kexin type 9 (PCSK9), is an enzyme which in humans is encoded by the PCSK9 gene.This gene encodes a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein plays a major regulatory role in cholesterol homeostasis. PCSK9 binds to the epidermal growth factor-like repeat A (EGF-A) domain of the low-density lipoprotein receptor (LDLR), inducing LDLR degradation. PCSK9 may also have a role in the differentiation of cortical neurons. Mutations in this gene have been associated with a rare form of autosomal dominant familial hypercholesterolemia (HCHOLA3).
This protein carries a polyhistidine tag at the C-terminus. This protein undergoes autocatalytic cleavage to release the pro-peptide and mature chain. The pro-peptide and mature chain are associated through non‐covalent interactions and with a calculated
Spécification
Spécification
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Rhesus macaque PCSK9, His Tag (PC9-C52H2) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession # A8T666).
-20°C
PCSK9
His Tag
HEK293
Rhesus macaque
13.9 kDa and 59.4 kDa
50 µg
Gln 31 - Gln 692
PCSK9,FH3,HCHOLA3,LDLCQ1,NARC1,PC9
Unconjugated
PBS, pH7.4