missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHOV Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26620-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The Rho subfamily of Ras-related GTPases controls multiple aspects of cell function, including cytoskeletal rearrangement, nuclear signaling and cell growth. Rho V (ras homolog gene family, member V), also known as Rho GTPase-like protein ARHV, CHP or WRCH2 (Wnt-1 responsive Cdc42 homolog 2), is a 236 amino acid protein that controls the actin cytoskeleton through activation of the JNK pathway. A member of the Rho family and small GTPase superfamily, Rho V functions as a lipid anchor at the cytoplasmic side of the cell membrane and is expressed in placenta, pancreas and fetal brain. Rho V is implicated in cell transformation and is encoded by a gene located on human chromosome 15, which houses over 700 genes and comprises nearly 3% of the human genome. Angelman syndrome, Prader-Willi syndrome, Tay-Sachs disease and Marfan syndrome are all associated with defects in chromosome 15-localized genes.Spécification
RHOV | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96L33, Q9Z1Y0 | |
ARHV, CDC42 like GTPase 2, Chp, GTP binding protein like 2, Rho GTPase like protein ARHV, RHOV, WRCH 2, WRCH2 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
171177, 171581 |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RHOV | |
Liquid | |
RHOV | |
RHOV Fusion Protein Ag24331 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Rat |