missing translation for 'onlineSavingsMsg'
Learn More

RNASEH2A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 16132-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16805654

  • 329.85€ / 150µL

Expédition estimée
02-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

YBX1 binds to splice sites in pre-mRNA and regulates splice site selection. This protein binds and stabilizes cytoplasmic mRNA and contributes to the regulation of translation by modulating the interaction between the mRNA and eukaryotic initiation factors. It binds to promoters that contain a Y-box (5′-CTGATTGGCCAA-3′), such as HLA class II genes. It regulates the transcription of numerous genes and promotes separation of DNA strands that contain mismatches or are modified by cisplatin. It has endonucleolytic activity and can introduce nicks or breaks into double-stranded DNA (in vitro), and it may play a role in DNA repair.
Spécification

Spécification

RNASEH2A
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75792
AGS4, JUNB, Ribonuclease H2 subunit A, ribonuclease H2, subunit A, Ribonuclease HI large subunit, Ribonuclease HI subunit A, RNase H(35), RNase H2 subunit A, RNase HI large subunit, RNASEH2A, RNASEHI, RNHIA, RNHL
Rabbit
IgG
150 μL
-20°C
Primary
10535
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RNASEH2A
Liquid
RNASEH2A
RNASEH2A Fusion Protein Ag9118
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Vidéos
FDS
Documents