missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RND2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13844-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The Ras p21 family of guanine nucleotide proteins has been widely studied in view of its apparent role in signal transduction pathways and high frequency of mutations in human maligcies. It is now clear, however, that the Ras proteins (H-, K- and N-Ras p21) are members of a much larger superfamily of related proteins. Six members of this family, Rap 1A, Rap 1B, Rap 2, R-Ras, Ral A and Ral B, exhibit approximately 50% amino acid homology to Ras. The six mammalian Rho proteins (Rho A, B, C, G, 7 and 8) are approximately 30% homologous to Ras and are expressed in a wide range of cell types. Both Ras p21 and Rho p21, as well as other members of the Ras superfamily, contain a carboxy-terminal CAAX sequence (C, cysteine; A, aliphatic amino acid; X, any amino acid) which in the case of Ras has been shown to be essential for correct localization and function.Spécification
RND2 | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P52198, Q9QYM5 | |
ARHN, Rho family GTPase 2, RHO7, RhoN, RND2 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
11858, 8153 |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
RND2 | |
Liquid | |
RND2 | |
RND2 Fusion Protein Ag4807 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse |