missing translation for 'onlineSavingsMsg'
Learn More

RNF141 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 26041-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16804575

  • 112.71€ / 20µL

Expédition estimée
02-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene contains a RING finger, a motif known to be involved in protein-DNA and protein-protein interactions. Abundant expression of this gene was found in the testicular tissue of fertile men, but was not detected in azoospermic patients. Studies of the mouse counterpart suggest that this gene may function as a testis specific transcription factor during spermatogenesis.
Spécification

Spécification

RNF141
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WVD5, Q99MB7
ring finger protein 141, RNF141, ZFP26, Zinc finger protein 230, ZNF230
Rabbit
IgG
20 μL
-20°C
Primary
50862, 67150
Immunoprecipitation, Western Blot
Unconjugated
RNF141
Liquid
RNF141
RNF141 Fusion Protein Ag23316
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Vidéos
FDS
Documents