missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF25 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24536-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene contains a RING finger motif. The mouse counterpart of this protein has been shown to interact with Rela, the p65 subunit of NF-kappaB (NFKB), and modulate NFKB-mediated transcription activity. The mouse protein also binds ubiquitin-conjugating enzymes (E2s) and is a substrate for E2-dependent ubiquitination.Spécification
RNF25 | |
Polyclonal | |
Unconjugated | |
RNF25 | |
AO7, ring finger protein 25, RNF25 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64320 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96BH1 | |
RNF25 | |
RNF25 Fusion Protein Ag21369 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |