missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPLP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21636-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. Ribosomal Protein LP1 (ribosomal protein, large, P1), also known as 60S acidic ribosomal protein P1, RPP1 or RPLP1, is a 114 amino acid acidic protein that belongs to the ribosomal protein L12P family. A key component during the elongation step of protein synthesis, Ribosomal Protein LP1 forms a pentameric complex (in conjunction with LP2 and LP0) that regulates ribosome assembly and plays a role in translation initiation. Like other mammalian ribosomal proteins, Ribosomal Protein LP1 exists as multiple processed pseudogenes that are found throughout the genome.Spécification
RPLP1 | |
Polyclonal | |
Unconjugated | |
RPLP1 | |
P1, ribosomal protein, large, P1, RPLP1, RPP1, RRP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6176 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P05386 | |
RPLP1 | |
RPLP1 Fusion Protein Ag16268 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |