missing translation for 'onlineSavingsMsg'
Learn More

RSPH9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 23253-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802265

  • 112.71€ / 20µL

Expédition estimée
03-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a protein thought to be a component of the radial spoke head in motile cilia and flagella. Mutations in this gene are associated with primary ciliary dyskinesia 12. Alternative splicing results in multiple transcript variants.
Spécification

Spécification

RSPH9
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9D9V4, Q9H1X1
C6orf206, CILD12, MRPS18AL1, RSPH9
Rabbit
IgG
20 μL
-20°C
Primary
221421, 316238, 75564
Western Blot
Unconjugated
RSPH9
Liquid
Rsph9
RSPH9 Fusion Protein Ag19745
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Rat, Human
Vidéos
FDS
Documents