missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSPO1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25348-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene is a member of the R-spondin family and encodes a secreted activator protein with two cystein-rich, furin-like domains and one thrombospondin type 1 domain. In mice, the protein induces the rapid onset of crypt cell proliferation and increases intestinal epithelial healing, providing a protective effect against chemotherapy-induced adverse effects.Spécification
RSPO1 | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q2MKA7 | |
CRISTIN3, hRspo1, R spondin 1, Roof plate specific spondin 1, RSPO, RSPO1 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
284654 |
Western Blot | |
Unconjugated | |
RSPO1 | |
Liquid | |
RSPO1 | |
RSPO1 Fusion Protein Ag21858 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |