missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RXRA Mouse anti-Human, Mouse, Clone: 4H6C4, Proteintech
Mouse Monoclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | RXRA |
---|---|
Clone | 4H6C4 |
Concentration | 0.45 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16847195
|
Proteintech
60198-1-IG-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16837195
|
Proteintech
60198-1-IG-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.Spécification
RXRA | |
0.45 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
NR2B1, Retinoid X receptor alpha, retinoid X receptor, alpha, RXRA | |
RXRA | |
IgG1 | |
Protein G | |
Antibody |
4H6C4 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
P19793, P28700 | |
20181, 6256 | |
RXRA Fusion Protein Ag0987 | |
Primary | |
-20°C | |
RXRA |