missing translation for 'onlineSavingsMsg'
Learn More

Acide (S)-2-(Fmoc-amino)butyrique, 98 %, Thermo Scientific™

Catalog Number 15433398
Click to view available options
Quantité:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
135112-27-5
C19H19NO4
325.364
MFCD00080268
XQIRYUNKLVPVRR-KRWDZBQOSA-N
fmoc-abu-oh, fmoc-2-abu-oh, fmoc-l-2-aminobutyric acid, fomc-abu-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-2-aminobutyric acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, butanoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, s, l-2-fmoc-amino butyric acid
853016
Acide (2S)-2-(9H-fluorène-9-ylméthoxycarbonylamino) butanoïque
CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
135112-27-5
C19H19NO4
325.364
MFCD00080268
XQIRYUNKLVPVRR-KRWDZBQOSA-N
fmoc-abu-oh, fmoc-2-abu-oh, fmoc-l-2-aminobutyric acid, fomc-abu-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino butanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-l-2-aminobutyric acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, butanoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, s, l-2-fmoc-amino butyric acid
853016
Acide (2S)-2-(9H-fluorène-9-ylméthoxycarbonylamino) butanoïque
CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau (S)-2-(Fmoc-amino)butyric acid
Quantité 1 g
Beilstein 7657232
Rotation optique −22° (c=1 in DMF)
Poids de la formule 325.36
Pourcentage de pureté 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'