missing translation for 'onlineSavingsMsg'
Learn More

S1PR5/EDG8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13874-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802284

  • 329.85€ / 150µL

Date d’expédition estimée: 18-06-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The lysosphingolipid sphingosine 1-phosphate (S1P) regulates cell proliferation, apoptosis, motility, and neurite retraction. Its actions may be both intracellular as a second messenger and extracellular as a receptor ligand. S1P and the structurally related lysolipid mediator lysophosphatidic acid (LPA) signal cells through a set of G protein-coupled receptors known as EDG receptors. Some EDG receptors (e.g., EDG1) are S1P receptors; others (e.g., EDG2) are LPA receptors.
Spécification

Spécification

S1PR5/EDG8
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q91X56, Q9H228, Q9JKM5
Edg 8, EDG8, S1P receptor 5, S1P receptor Edg 8, S1P5, S1PR5, SPPR 1, SPPR 2
Rabbit
IgG
150 μL
-20°C
Primary
53637, 60399, 94226
Flow Cytometry, Western Blot
Unconjugated
S1PR5
Liquid
S1PR5
S1PR5/EDG8 Fusion Protein Ag4928
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Vidéos
FDS
Documents