missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10229-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Posttranslational modification of proteins by the addition of the small protein SUMO (see SUMO1), or sumoylation, regulates protein structure and intracellular localization. SAE1 and UBA2. form a heterodimer that functions as a SUMO-activating enzyme for the sumoylation of proteins (Okuma et al., 1999).Spécification
SAE1 | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6AXQ0, Q9R1T2, Q9UBE0 | |
AOS1, FLJ3091, HSPC140, SAE1, SUA1, UBLE1A | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
10055, 308384, 56459 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SAE1 | |
Liquid | |
Sae1 | |
SAE1 Fusion Protein Ag0275 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |