missing translation for 'onlineSavingsMsg'
Learn More

SALL2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 23114-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802205

  • 112.71€ / 20µL

Expédition estimée
09-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody is specific to the SALL2 protein.

This gene encodes a protein containing multiple zinc finger domains. The encoded protein functions in optical fissure closure during development of the eye in the embryo. Mutations in this gene are associated with ocular coloboma.
Spécification

Spécification

SALL2
0.57 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y467
HSAL2, KIAA0360, p150(Sal2), Sal 2, sal like 2 (Drosophila), Sal like protein 2, SAL2, SALL2, Zinc finger protein SALL2, Zinc finger protein Spalt 2, ZNF795
Rabbit
IgG
20 μL
-20°C
Primary
6297
Western Blot
Unconjugated
SALL2
Liquid
SALL2
SALL2 Fusion Protein Ag19427
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Vidéos
FDS
Documents