missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMD11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24731-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
SAMD11 | |
Polyclonal | |
Unconjugated | |
SAMD11 | |
SAMD11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
148398 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96NU1 | |
SAMD11 | |
SAMD11 Fusion Protein Ag20671 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |