Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S protein RBD (N354D), His Tag
Marque:  ACROBiosystems SPD-S52H5-1mg
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16625332
  • 2622.86€ / 1mg
Date d’expédition estimée: 01-12-2021
Description
Description
It's been reported that SARS-CoV-2 can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.0 kDa. The protein migrates as 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification
Spécification
QHD43416.1 (N354D)
HEK293
SARS-CoV-2
27.0 kDa
1 mg
Arg 319 - Phe 541
Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Viral Proteins
Powder
>95% as determined by SDS-PAGE
SARS-CoV-2 S protein RBD (N354D), His Tag (SPD-S52H5) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Phe 541 (Accession # QHD43416.1 (N354D)).
-20°C
S protein RBD
His Tag