Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S1 protein (Y144del), His Tag
Marque:  ACROBiosystems S1N-C52Hc-1mg
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 17256460
  • 2838.77€ / 1mg
Expédition estimée: 26-07-2022
pour voir le stock disponible 
Description
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 76.7 kDa. The protein migrates as 100-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification
Spécification
QHD43416.1 (Y144del)
>95% as determined by SDS-PAGE.
SARS-CoV-2 S1 protein (Y144del), His Tag (S1N-C52Hc) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1 (Y144del)). Y144 deletion was identified in the SARS-CoV-2 variant (known as 20 B/501 Y.V1, VOC 202012/01, or B0.1.1.7 lineage) which emerged in the United Kindom.
-20°C
S1 protein
His Tag
Less than 1.0 EU / µg by the LAL method.
Powder
SARS-CoV-2
76.7 kDa
1 mg
Val 16 - Arg 685
Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1,COVID-19
Unconjugated
PBS, pH7.4
HEK293
Viral Proteins