Learn More
ACROBiosystems SARS-CoV-2 Nucleocapsid protein (D3L, R203K, G204R, S235F), His Tag
483.87€ - 3495.90€
Spécification
Numéro d’adhésion | QHO62115.1 |
---|---|
Poids moléculaire | 47.4 kDa |
Source | SARS-CoV-2 Nucleocapsid protein, His Tag (NUN-C52H8) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1). D3L/ R203K/ G204R/ S235F were identified in the SARS-CoV-2 variant (known as 20 B/501 Y.V1, VOC 202012/01, or B0.1.1.7 lineage) which emerged in the United Kindom. |
Immunogène | Met 1 - Ala 419 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
17276360
|
ACROBiosystems
NUN-C52H8-1MG |
1 mg |
3495.90€
1mg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
17266360
|
ACROBiosystems
NUN-C52H8-100UG |
100 μg |
483.87€
100µg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Nucleocapsid (N) protein is the most abundant protein found in coronavirus. CoV N protein is a highly immunogenic phosphoprotein important for viral genome replication and modulation of cell signaling pathways. It was first identified by a research team while they were screening for ADP-ribosylated proteins during coronavirus (CoV) infection (Grunewald M. E., et al. 2017, Virology; 517: 62-68). The array of diverse functional activities accommodated in N protein makes it more than a structural protein but also an interesting target in the development of antiviral therapeutics. Because of the conservation of N protein sequence and its strong immunogenicity, N protein of coronavirus is chosen as a diagnostic tool.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.4 kDa. The protein migrates as 30-35 kDa and 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
QHO62115.1 | |
SARS-CoV-2 Nucleocapsid protein, His Tag (NUN-C52H8) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1). D3L/ R203K/ G204R/ S235F were identified in the SARS-CoV-2 variant (known as 20 B/501 Y.V1, VOC 202012/01, or B0.1.1.7 lineage) which emerged in the United Kindom. | |
-20°C | |
Nucleocapsid protein | |
SARS-CoV-2 | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |
47.4 kDa | |
Met 1 - Ala 419 | |
Nucleocapsid protein,NP,Protein N,COVID-19 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE. |