missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems SARS-CoV-2 S1 protein NTD (A262S), His Tag (MALS verified)
387.09€ - 2782.20€
Spécification
Numéro d’adhésion | QHD43416.1 (A262S) |
---|---|
Poids moléculaire | 34.9 kDa |
Source | SARS-CoV-2 S1 protein NTD (A262S), His Tag (S1D-C52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303 (Accession # QHD43416.1 (A262S)). |
Immunogène | Ser 13 - Leu 303 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
17276440
|
ACROBiosystems
S1D-C52H5-1MG |
1 mg |
2782.20€
1mg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
17266440
|
ACROBiosystems
S1D-C52H5-100UG |
100 μg |
387.09€
100µg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.9 kDa. The protein migrates as 45-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
QHD43416.1 (A262S) | |
SARS-CoV-2 S1 protein NTD (A262S), His Tag (S1D-C52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303 (Accession # QHD43416.1 (A262S)). | |
-20°C | |
S1 protein NTD | |
SARS-CoV-2 | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |
34.9 kDa | |
Ser 13 - Leu 303 | |
S1 protein NTD,Spike protein S1 NTD,BetaCoV S1-NTD,COVID-19 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE. |