missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCNN1G Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13943-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Epithelial sodium channels are amiloride-sensitive members of the degenerin/epithelial sodium channel (Deg/ENaC) superfamily of ion channels. Members of this superfamily of ion channels share organizational similarity in that they all possess two short intracellular amino and carboxyl termini, two short membrane spanning segments, and a large extracellular loop with a conserved cysteine-rich region. There are three homologous isoforms of the ENaC (alpha, beta, and gamma) protein. ENaC in the kidney, lung, and colon plays an essential role in trans-epithelial sodium and fluid balance. ENaC also mediates aldosterone-dependent sodium reabsorption in the distal nephron of the kidney, thus regulating blood pressure. ENaC is thought to be regulated, in part, through association with the cystic fibrosis transmembrane conductance regulator (CFTR) chloride ion channel. Gain-of-function mutations in beta- or gamma-ENaC can cause severe arterial hypertension (Liddel′s syndrome) and loss-of-function mutations in alpha- or beta-ENaC causes pseudohypoaldosteronism (PHA-1).Spécification
SCNN1G | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P37091, P51170, Q9WU39 | |
ENaCg, ENaCgamma, Gamma ENaC, Gamma NaCH, PHA1, SCNEG, SCNN1G | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
20278, 24768, 6340 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SCNN1G | |
Liquid | |
SCNN1G | |
SCNN1G Fusion Protein Ag5020 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |