missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCRN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14303-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene likely encodes a member of the secernin family of proteins. A similar protein in rat functions in regulation of exocytosis in mast cells. Alternatively spliced transcript variants have been described.Spécification
SCRN1 | |
Polyclonal | |
Unconjugated | |
SCRN1 | |
KIAA0193, SCRN1, secernin 1, SES1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
502776, 69938, 9805 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q12765, Q6AY84, Q9CZC8 | |
Scrn1 | |
SCRN1 Fusion Protein Ag5413 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |