missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC11A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14753-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the peptidase S26B family. The encoded protein is an 18kDa subunit of the signal peptidase complex and has been linked to cell migration and invasion, gastric cancer and lymph node metastasis. Alternative splicing results in multiple transcript variants. A related pseudogene has been identified on chromosome 8.Spécification
SEC11A | |
Polyclonal | |
Unconjugated | |
SEC11A | |
1810012E07Rik, Endopeptidase SP18, SEC11 homolog A, SEC11 like protein 1, SEC11A, SEC11L1, sid2895, SPase 18 kDa subunit, SPC18, SPCS4A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23478, 56529, 65166 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P42667, P67812, Q9R0P6 | |
Sec11a | |
SEC11A Fusion Protein Ag6500 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |