missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC24A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15958-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
SC24A is a component of coat protein II (COPII; see MIM 601924)-coated vesicles that mediate protein transport from the endoplasmic reticulum. SEC24A is 1 of several mammalian proteins that show structural and functional homology to yeast Sec24.Spécification
SEC24A | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95486, Q3U2P1 | |
SEC24 related protein A, SEC24A | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
10802, 287275, 77371 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SEC24A | |
Liquid | |
Sec24a | |
SEC24A Fusion Protein Ag8723 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |