missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC61B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14846-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The Sec61 complex is responsible for translocation of proteins across the rough endoplasmic reticulum membrane. Sec61 is composed of three subunits: alpha, beta, and gamma; that associate to form a heterotrimer. Multiple heterotrimers associate forming the ER translocon. Sec61 alpha is a polytopic protein that has ten membrane-spanning loops. Sec61 has been found to be tightly associated with membrane bound ribosomes, either directly or through adaptor proteins, and is required for assembly of membrane and secretory proteins.Spécification
SEC61B | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P60468, Q9CQS8 | |
Sec61 beta subunit, SEC61B | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
10952, 66212 |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
SEC61B | |
Liquid | |
Sec61b | |
Peptide | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Human |