missing translation for 'onlineSavingsMsg'
Learn More

Sendai virus HN Mouse anti-Virus, Alexa Fluor 488, Clone: 1A6, Invitrogen™

Mouse Monoclonal Antibody

Marque:  Affymetrix eBioscience 53-6494-80

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 16636414

  • 154.00€ / 2 pièces

Date d’expédition estimée: 29-10-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Description: This 1A6 monoclonal antibody recognizes hemagglutinin-neuraminidase of Sendai virus. This clone has been successfully used in flow cytometry, immunocytochemistry and has been reported for neutralization. The CTS CytoTune-iPS 2.1 Sendai Reprogramming Kit and the CytoTune-iPS 2.0 Sendai Reprograming Kit utilize Sendai virus particles to deliver Yamanaka factors into target cells in order to reprogram them into induced Pluripotent Stem Cells (iPSC). Clone 1A6 can be used to detect the presence of the residual Sendai virus after transduction and subsequent tissue culture. Applications Reported: This 1A6 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1A6 antibody has been tested by flow cytometric analysis of Sendai virus-infected cells. This may be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser

Sendai virus is a single stranded RNA respirovirus from the family Paramyxoviridae, which replicates in the respiratory tract and infects certain mammals, including mice, hamsters, rats and guinea pigs. The virus is highly contagious and in the wild is transmitted through contact with aerosolized respiratory secretions. The Hemagglutinin-Neuraminidase (HN) protein of the Sendai virus facilitates cell entry and infection by binding and cleaving terminal sialic acids present on many cell surface glycoproteins.
Spécification

Spécification

Sendai virus HN
1A6
Alexa Fluor 488
Liquid
Sendai Virus
Affinity chromatography
RUO
Antibody
Monoclonal
Flow Cytometry
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Mouse
IgG2a, kappa
25 μg
4°C, store in dark, DO NOT FREEZE!
Primary
Virus
Vidéos
FDS
Documents
Certificats