Learn More
SFRP4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15328-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody got 41-51 kDa and 55 kDa bands in western blotting maybe due to differential posttranslational protein modification.
Secreted frizzled-related protein 4 (SFRP4) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. The expression of SFRP4 in ventricular myocardium correlates with apoptosis related gene expression.Spécification
SFRP4 | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6FHJ7 | |
FRP 4, FRPHE, sFRP 4, SFRP4 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
6424 |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
SFRP4 | |
Liquid | |
SFRP4 | |
SFRP4 Fusion Protein Ag6909 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |