missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGSH Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14184-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
SGSH is one of several enzymes involved in the lysosomal degradation of heparan sulfate. Mutations in this gene are associated with Sanfilippo syndrome A, one type of the lysosomal storage disease mucopolysaccaridosis III, which results from impaired degradation of heparan sulfate. Transcripts of varying sizes have been reported but their biological validity has not been determined.Spécification
SGSH | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P51688 | |
HSS, MPS3A, SFMD, SGSH, Sulfoglucosamine sulfamidase, Sulphamidase | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
6448 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SGSH | |
Liquid | |
SGSH | |
SGSH Fusion Protein Ag5435 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |