missing translation for 'onlineSavingsMsg'
Learn More

CSF1 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the N-terminal fragment (Met 1-Asn 190) of human CSF1 (P09603-1) fused to a polyhistidine tag at the C-terminus

Marque:  Invitrogen 11792H08H50

Code nomenclature Nacres: NA.28

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 13473059

  • 626.28€ / 5 pièces


Pour délai, nous consulter 03 88 67 14 14
Ajouter au panier

Description

Description

Macrophage Colony Stimulating Factor (CSF1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CSF1R (100μL/well of a 10μg/mL solution) can bind biotinylated human CSF1; the EC50 of biotinylated human CSF1 is 8ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Spécification

Spécification

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
19.8kDa
50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Colony stimulating factor 1 (macrophage)
Assay Standard
Mammalian
Lyophilized
>98% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Colony Stimulating Factors
RP11-195M16.2, MCSF, mgC31930
CSF1
Cytokines & Receptors
His Tag
Vidéos

For Research Use Only. Not for use in diagnostic procedures.