missing translation for 'onlineSavingsMsg'
Learn More

FCGR2b (CD32) Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 217) of mouse FCGR2b precursor (NP_001070657.1) fused to a polyhistidine tag at the C-terminus

Marque:  Invitrogen 50030M08H5

Code nomenclature Nacres: NA.28

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 13475616

  • 68.25€ / 5µg

Expédition estimée
19-08-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Fc gamma receptor II-b (FCGR2b/CD32), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature.

Spécification

Spécification

Mammalian
Glycoproteins|Fc Receptors
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
14130
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Fc receptor, IgG, low affinity IIb
His Tag
Growth Factors & Receptors/CD Proteins
Assay Standard
Lyophilized
21.7kDa
5μg
Truncated/Recombinant/Cell Surface Protein
AI528646, CD32, F630109E10Rik, Fcgr2, Fcgr2a, Fcr-2, Fcr-3, Ly-17, Ly-m20, Lym-1, FCGR2b
FCGR2b
Vidéos
Documents

For Research Use Only. Not for use in diagnostic procedures.