missing translation for 'onlineSavingsMsg'
Learn More

FCGR3B (CD16b) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ser 200) of human FCGR3B (NP_000561.3) fused to a polyhistidine tag at the C-terminus

Marque:  Invitrogen 11046H08H50

Code nomenclature Nacres: NA.28

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 13476036

  • 250.00€ / 50µg

Expédition estimée
24-08-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Fc gamma receptor III-b (FCGR3B/CD16b), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Spécification

Spécification

Mammalian
Glycoproteins|Fc Receptors
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
2215
≥95% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Fc fragment of IgG, low affinity IIIb, receptor (CD16b)
His Tag
CD Proteins
Assay Standard
Lyophilized
22.2kDa
50μg
Truncated/Recombinant/Cell Surface Protein
CD16, CD16b, FCG3, FCGR3, FCGR3B
FCGR3B
Vidéos
Documents

For Research Use Only. Not for use in diagnostic procedures.