missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLAIN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22123-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
SLAIN1 (SLAIN Motif Family Member 1) is a Protein Coding gene. An important paralog of this gene is SLAIN2. Microtubule plus-end tracking protein that might be involved in the regulation of cytoplasmic microtubule dynamics, microtubule organization and microtubule elongation.Spécification
SLAIN1 | |
Polyclonal | |
Unconjugated | |
SLAIN1 | |
C13orf32, SLAIN motif family, member 1, SLAIN1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
105439, 122060 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q68FF7, Q8ND83 | |
Slain1 | |
SLAIN1 Fusion Protein Ag4973 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |