missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SLAMF9 Synthetic Peptide
Synthetic Peptide
Marque: Invitrogen™ PEP1217
Code nomenclature Nacres: NA.26
Informations supplémentaires : Poids : 0.01000kg
Description
SLAMF9 Synthetic Peptide, PEP-1217, from Invitrogen.
This peptide corresponds to 19 amino acid peptide near the carboxy terminus of human SLAMF9.PEP-1217 can be used as a blocking peptide with polyclonal antibody PA5-21103.Spécification
89886 | |
purified | |
Human SLAMF9 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q96A28 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
SLAMF9 | |
0.02% sodium azide | |
50 μg | |
PBS |