missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC12A4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15927-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
SLC12A4 mediates electroneutral potassium-chloride cotransport when activated by cell swelling. SLC12A4 may contribute to cell volume homeostasis in single cells.Spécification
SLC12A4 | |
Polyclonal | |
Unconjugated | |
SLC12A4 | |
Erythroid K Cl cotransporter 1, hKCC1, KCC1, SLC12A4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20498, 29501, 6560 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q63632, Q9JIS8, Q9UP95 | |
SLC12A4 | |
SLC12A4 Fusion Protein Ag8762 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |