Learn More
SLC16A4 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 20889-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16830475
  • 121.38€ / 20µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 
Description
Description
The SLC16A4 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SLC16A4. This antibody recognizes human antigen. The SLC16A4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Spécification
SLC16A4
Polyclonal
Unconjugated
20 μL
9122
IgG
ELISA, Immunohistochemistry, Western Blot
280 μg/mL