missing translation for 'onlineSavingsMsg'
Learn More

SLC26A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 27759-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16805815

  • 112.71€ / 20µL

Expédition estimée
08-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The SLC26 family is comprised of sulfate/anion transporters that are well conserved in both their genomic and protein structures, yet have markedly different tissue expression patterns. Members of the SLC26 family can mediate the electroneutral exchange of Cl- for HCO3- across the plasma membrane of mammalian cells. SLC26A2 (solute carrier family 26, member 2), also known as DTD, EDM4, DTDST, MST153, D5S1708 or MSTP157, is a 739 amino acid diastrophic dysplasia sulfate transporter belonging to the SLC26A/SulP transporter family. Ubiquitously expressed, SLC26A2 may play a role in endochondral bone formation. SLC26A2 is a transmembrane glycoprotein implicated in the pathogenesis of several human chondrodysplasias and is critical for sulfation of proteoglycans and matrix organization in cartilage.
Spécification

Spécification

SLC26A2
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O70531, P50443, Q62273
D5S1708, Diastrophic dysplasia protein, DTD, DTDST, EDM4, MST153, MSTP157, SLC26A2, Sulfate transporter
Rabbit
IgG
20 μL
-20°C
Primary
117267, 13521, 1836
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
SLC26A2
Liquid
SLC26A2
SLC26A2 Fusion Protein Ag26521
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Vidéos
FDS
Documents