missing translation for 'onlineSavingsMsg'
Learn More

SLC39A7 Recombinant Protein (P01), Abnova

Marque:  Abnova H00007922-P01.25ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16122202

  • 467.00€ / 25µg

Expédition estimée
21-12-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

AAH00645
Solution
7922
SLC39A7 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SLC39A7
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
73.7
8
Glutathione Sepharose 4 Fast Flow
25 ug
DDLHDDLQEDFHGHSHRHSHEDFHHGHSHAHGHGHTHESIWHGHTHDHDHGHSHEDLHHGHSHGYSHESLYHRGHGHDHEHSHGGYGESGAPGIKQDLDAVTLWAYALGATVLISAAPFFVLFLIPVESNSPRHRSLLQILLSFASGGLLGDAFLHLIPHALEPHSHHTLEQPGHGHSHSGQGPILSVGLWVLSGIVAFLVVEKFVRHVKGGHGHSHGHGHAHSHTRGSHGHGRQERSTKEKQSSEEEEKETRGVQKRRGGSTVPKDGPVRPQNAEEEKRGLDLRVSGYLNLAADLAHNFTDGLAIGASFRGGRGLGILTTMTVLLHEVPHEVGDFAILVQSGCSKKQAMRLQLLTAVGALAGTACALLTEGGAVGSEIAGGAGPGWVLPFTAGGFIYVATVSVLPELLREASPLQSLLEVLGLLGGVIMMVLIAHLE
D6S115E|D6S2244E|H2-KE4|HKE4|KE4|RING5|ZIP7
SLC39A7
Wheat Germ (in vitro)
GST
Vidéos
FDS
Documents