missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC7A5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13752-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Solute carrier family 7 member 5 also known as large neutral amino acids transporter small subunit 1 is a protein that in humans is encoded by the SLC7A5 gene.It is sodium-independent, high-affinity transport of large neutral amino acids such as phenylalanine, tyrosine, leucine, arginine and tryptophan, when associated with SLC3A2/4F2hc.It involved in cellular amino acid uptake and the transport of L-DOPA across the blood-brain barrier, and that of thyroid hormones triiodothyronine (T3) and thyroxine (T4) across the cell membrane in tissues such as placenta.Acts as an amino acid exchanger and plays a role in neuronal cell proliferation (neurogenesis) in brain.Spécification
SLC7A5 | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q01650, Q63016, Q9Z127 | |
4F2 light chain, 4F2LC, CD98 light chain, CD98LC, D16S469E, E16, hLAT1, Integral membrane protein E16, LAT1, MPE16, SLC7A5 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
20539, 50719, 8140 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SLC7A5 | |
Liquid | |
Slc7a5 | |
SLC7A5 Fusion Protein Ag4694 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Human, Rat |