missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLCO4C1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24584-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Organic anion transporter, capable of transporting pharmacological substances such as digoxin, ouabain, thyroxine, methotrexate and cAMP. May participate in the regulation of membrane transport of ouabain. Involved in the uptake of the dipeptidyl peptidase-4 inhibitor sitagliptin and hence may play a role in its transport into and out of renal proximal tubule cells. May be involved in the first step of the transport pathway of digoxin and various compounds into the urine in the kidney. May be involved in sperm maturation by enabling directed movement of organic anions and compounds within or between cells. This ion- transporting process is important to maintain the strict epididymal homeostasis necessary for sperm maturation. May have a role in secretory functions since seminal vesicle epithelial cells are assumed to secrete proteins involved in decapacitation by modifying surface proteins to facilitate the acquisition of the ability to fertilize the egg.Spécification
SLCO4C1 | |
Polyclonal | |
Unconjugated | |
SLCO4C1 | |
OATP H, OATP M1, OATP4C1, OATPX, Organic anion transporter M1, PRO2176, SLC21A20, SLCO4C1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
227394, 353189 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6ZQN7, Q8BGD4 | |
SLCO4C1 | |
SLCO4C1 Fusion Protein Ag20177 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |