Learn More
Cole-Parmer™ CHRONOMETRE
Easy to hold and features large controls
Marque: Cole-Parmer™ 94460-00
Code nomenclature Nacres: NB.51
Informations supplémentaires : Poids : 0.80000kg
Description
- Get accurate, reliable measurements with included NIST-traceable calibration
- Counts down from 100 minutes in 1-second intervals
- A 30-second audible alarm sounds when time has elapsed
- calibrated to NIST-traceable standards and ready to use
Spécification
Digital Timer | |
±8.64sec./day | |
Digital LCD | |
7.93 mm H | |
1 Each |
100 min. | |
1 sec. | |
1 | |
Magnetic Back, one 1.5 V Button Cell Battery, and NIST-Traceable Calibration Report Supplied By the Manufacturer | |
NIST-Traceable Calibration Report Supplied By The Manufacturer |