Learn More
SMCR7/MID49 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28718-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The SMCR7/MID49 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SMCR7/MID49. This antibody recognizes human, mouse, rat antigen. The SMCR7/MID49 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
SMCR7/MID49 | |
Polyclonal | |
Unconjugated | |
20 μL | |
125170 | |
IgG |
Western Blot, Immunohistochemistry | |
467 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |