missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMPDL3B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16552-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Located on chromosome 1, this gene encodes for acid sphingomyelinase-like phosphodiesterase 3b precursor protein.Spécification
SMPDL3B | |
Polyclonal | |
Unconjugated | |
SMPDL3B | |
ASM like phosphodiesterase 3b, ASML3B, SMPDL3B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100340, 27293, 362619 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P58242, Q92485 | |
SMPDL3B | |
SMPDL3B Fusion Protein Ag9535 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |