missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14789-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene belongs to the small nuclear ribonucleoprotein core protein family. It is required for pre-mRNA splicing and small nuclear ribonucleoprotein biogenesis. Multiple transcript variants encoding different isoforms have been found for this gene.Spécification
SNRPD2 | |
Polyclonal | |
Unconjugated | |
SNRPD2 | |
Sm D2, SMD2, snRNP core protein D2, SNRPD2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
107686, 6633, 680309 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62316, P62317 | |
SNRPD2 | |
SNRPD2 Fusion Protein Ag6443 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |