missing translation for 'onlineSavingsMsg'
Learn More

SNX15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 16049-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16805524

  • 329.85€ / 150µL

Expédition estimée
03-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a member of the sorting nexin family. Members of this family contain a phox domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. Overexpression of this gene results in a decrease in the processing of insulin and hepatocyte growth factor receptors to their mature subunits. This decrease is caused by the mislocalization of furin, the endoprotease responsible for cleavage of insulin and hepatocyte growth factor receptors. This protein is involved in endosomal trafficking from the plasma membrane to recycling endosomes or the trans-Golgi network. Alternative splicing results in multiple transcript variants. Read-through transcription also exists between this gene and the upstream ADP-ribosylation factor-like 2 gene.
Spécification

Spécification

SNX15
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4V896, Q91WE1, Q9NRS6
HSAF001435, SNX15, sorting nexin 15
Rabbit
IgG
150 μL
-20°C
Primary
293691, 29907, 69024
Western Blot
Unconjugated
SNX15
Liquid
Snx15
SNX15 Fusion Protein Ag9006
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Vidéos
FDS
Documents