missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX27 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16329-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
SNX27 encodes a member of the sorting nexin family, a diverse group of cytoplasmic and membrane-associated proteins involved in endocytosis of plasma membrane receptors and protein trafficking through these compartments. All members of this protein family contain a phosphoinositide binding domain (PX domain). A highly similar protein in mouse is responsible for the specific recruitment of an isoform of serotonin 5-hydroxytryptamine 4 receptor into early endosomes, suggesting the analogous role for the human protein.Spécification
SNX27 | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3UHD6, Q8K4V4, Q96L92 | |
KIAA0488, MRT1, MY014, SNX27, Sorting nexin 27, sorting nexin family member 27 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
260323, 76742, 81609 |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
SNX27 | |
Liquid | |
Snx27 | |
SNX27 Fusion Protein Ag9447 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Rat, Human |