missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Sodium carbonate, Free-flowing, ACS reagent, anhydrous, ≥99.5%, Honeywell Fluka™

Free-flowing, ACS reagent, anhydrous,≥99.5%

Marque:  Honeywell Fluka™ 791768-12KG

Code nomenclature Nacres: NA.21

Informations supplémentaires : CAS : 497-19-8 Poids : 12.00000kg

Quantité. 12kg
Emballage Fiberboard Box

 Afficher plus de versions de ce produit

Code produit. 15657090

  • 480.10€ / 12kg


En stock 10
Ajouter au panier

Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spécification

Identifiants chimiques

497-19-8
105.988
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
CHEBI:29377
C(=O)([O-])[O-].[Na+].[Na+]
CNa2O3
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
disodium;carbonate

Spécification

Sodium carbonate
497-19-8
≤100.0000%
CNa2O3
4154566
CDBYLPFSWZWCQE-UHFFFAOYSA-L
disodium;carbonate
10340
105.99
>99.50%
2.53 g/cm3
851°C (literature)
Free-flowing, ACS reagent, anhydrous, ≥99.5%
≥90.0000%
calc. to the dried substance
Na2CO3
sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
C(=O)([O-])[O-].[Na+].[Na+]
105.988
CHEBI:29377
anhydrous
ACS Reagent
Fiberboard Box
12kg
Vidéos
FDS